missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MMP10 ELISA Kit DFS Item

ELISA

$866.00 - $866.00

Specifications

Accession Number P09238
Assay Range 2.06 to 1500 pg/mL
Assay Sensitivity 3 pg/mL
Calibration Range 3 pg/mL to 1500 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
EHMMP10 DFS Item
View Documents
Invitrogen™
EHMMP10
96 Tests N/A
Only null left
 
Description

Description

The Human Matrix MetalloProteinase 10 (Hu MMP10) ELISA quantitates Hu MMP10 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MMP10. Principle of the method The Human MMP10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.
Specifications

Specifications

P09238
3 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
SL-2,STMY2
<10
HRP
Extracellular Matrix & Cell Adhesion
2°C to 8°C
MMP10
4 hrs 45 mins
2.06 to 1500 pg/mL
3 pg/mL to 1500 pg/mL
HRP
Cell Structure & Organelle Proteins
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, EGF, ENA-78, FGF-4, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, 6, 7, 9, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, MMP-1, -2, -3, -9,- 12,-13, PARC, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
4319
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
SL-2, STMY2
1 hrs 20 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'