missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP1 Monoclonal Antibody (K29028_6B4), Invitrogen™
Mouse Monoclonal Antibody
$1,557.73 - $2,745.56
Specifications
| Antigen | MMP1 |
|---|---|
| Clone | K29028_6B4 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: QTHLTYRIEN YTPDLPRADV DHAIEKAFQL WSNVTPLTFT KVSEGQADIM ISFVRGDHRD NSPFDGPGGN LAHAFQPGPG IGGDAHFDED ERWTNNFREY NLHRVAAHEL GHSLGLSHST DIGALMYPSY TFSGDVQLAQ DDIDGIQAIY GRSQNPVQPI GPQTPKACDS KLTFDAITTI
MMP1 Monoclonal antibody specifically detects MMP1 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| MMP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1a; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1a; MMPs | |
| Recombinant human MMP1 (immunogen range is 111-290). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MMP1 |
| K29028_6B4 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P03956 | |
| 4312 | |
| IgG2a | |
| Protein A | |
| Antibody |