missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (23GB2935)

Catalog No. MA553519
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553519 20 μL
MA553518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553519 Supplier Invitrogen™ Supplier No. MA553519
Only null left

Rabbit Recombinant Monoclonal Antibody

MLKL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.

Specifications

Antigen MLKL
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2935
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mlkl
Gene Accession No. Q8NB16
Gene Alias 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Gene Symbols Mlkl
Host Species Rabbit
Immunogen A synthesized peptide derived from MLKL (1-200AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less