missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLANA Mouse anti-Human, Clone:2F3B7, Abnova™
Mouse monoclonal antibody raised against recombinant human MLANA.
Supplier: Abnova Corporation MAB16603
Specifications
| MLANA | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| MLANA | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 2F3B7 | |
| melan-A | |
| In PBS (0.05% sodium azide) | |
| MART-1, MART1 | |
| Recombinant protein corresponding to amino acid 48-118 of human MLANA from E. coli. | |
| 2315 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |