missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ D-Tipper™ Pipetters

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Color coded body for easy volume identification and individually serialized with Calibration Certificate included.

Supplier:  VistaLab™ Technologies 1053C

 View more versions of this product

Catalog No. 21-354


Only null left
Explore available promotions

Description

Description

  • Single-stroke 'to deliver' operation
  • Built-in tip-ejector and glove guard
  • Factory calibrated to deliver specific quantities; can be easily recalibrated in the lab
  • Available in 30 fixed volumes ranging from 5-1000 μL
  • Also available as a Selectable model: 50/100/200 μL
Specifications

Specifications

±3%
White
Aluminum, Stainless Steel
1
Axial Pipette
Micro Pipetting/Liquid Transfer
1.12%
Not Autoclavable
Reusable
D-Tipper
Non-sterile
25 μL
Nozzle Insert, Calibration Key
Limited Lifetime
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions