missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ D-Tipper™ Pipetters

Catalog No. 2131915
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
30 μL
50 μL
100 μL
200 μL
250 μL
300 μL
400 μL
500 μL
1000 μL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
2131915 30 μL
2131925 400 μL
21354 25 μL
21360 200 μL
21362 250 μL
21364 300 μL
21381500 10 μL
21381502 20 μL
21381504 50 μL
21381506 100 μL
21381508 500 μL
21381510 1000 μL
21381515 5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 2131915 Supplier VistaLab™ Technologies Supplier No. 1143C
Only null left

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Color coded body for easy volume identification and individually serialized with Calibration Certificate included.

  • Single-stroke 'to deliver' operation
  • Built-in tip-ejector and glove guard
  • Factory calibrated to deliver specific quantities; can be easily recalibrated in the lab
  • Available in 30 fixed volumes ranging from 5-1000 μL
  • Also available as a Selectable model: 50/100/200 μL

Specifications

Accuracy ±3%
Autoclavable Not Autoclavable
Color Silver
Disposable Reusable
Material Aluminum, Stainless Steel
Model D-Tipper
Number of Channels 1
Sterility Non-sterile
Type Axial Pipette
Volume (Metric) 30 μL
For Use With (Application) Micro Pipetting/Liquid Transfer
Includes Nozzle Insert, Calibration Key
Precision 0.93%
Warranty Limited Lifetime
Show More Show Less