missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MKX Monoclonal Antibody (OTI6A12)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526996

Catalog No. PIMA526996


Only null left

Description

Description

MKX Monoclonal Antibody for Western Blot, IHC (P)

MKX is a member of an Iroquois (IRX) family-related class of 'three-amino acid loop extension' (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.
Specifications

Chemical Identifiers

MKX
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYA7
283078
Full length human recombinant protein of MKX produced in E.coli.
Antibody
OTI6A12
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG1
Human
MKX
9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652
MKX
Primary
Affinity Chromatography

Specifications

MKX
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYA7
MKX
Full length human recombinant protein of MKX produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI6A12
Unconjugated
MKX
9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652
Mouse
Affinity Chromatography
RUO
283078
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions