missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MKNK2 (MNK2), GST Tag Recombinant Protein

Catalog No. pv5608
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5608 1 mg
PV5607 10 μg
PR8046A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV5608 Supplier Invitrogen™ Supplier No. PV5608

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

The Map kinase-interacting kinases (MKNK1 and MKNK2) are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis. Overexpression of Mnk2 results in increased phosphorylation of endogenous eIF-4E, showing that it can act as an eIF-4E kinase in vivo. Mnk2 may play a role in the response to environmental stress and cytokines. Expression of active mutants of MNK1 and MNK2 in 293 cells diminishes cap-dependent translation relative to cap-independent translation in a transient reporter assay. Human Mnk2 is homologous to murine Mnk2 (approximately 94% identical) and human Mnk1 (71% identical). In vitro phosphorylation studies show that Mnk2 is a significantly better substrate than Mnk1 for extracellular signal-regulated kinase 2 (Erk2), p38MAPKalpha, and p38MAPKbeta.

Specifications

Accession Number Q9HBH9
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 2872
Molecular Weight (g/mol) 74.2 kDa
Name Human MKNK2 (MNK2), GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias 2010016G11Rik; G protein-coupled receptor kinase 7; GPRK7; MAP kinase interacting serine/threonine kinase 2; MAP kinase signal-integrating kinase 2; MAP kinase-interacting serine/threonine kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; LOW QUALITY PROTEIN: MAP kinase-interacting serine/threonine-protein kinase 2; MAPK interacting serine/threonine kinase 2; MAPK signal-integrating kinase 2; Mknk2; MNK2
Common Name MKNK2
Kinase Family MAPKAPK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less