missing translation for 'onlineSavingsMsg'
Learn More

Mixed Multi-Element Calibration Standards, SPEX CertiPrep™

Catalog No. mxstdsetan
Change view
Click to view available options
Quantity:
5 x 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MXSTDSETAN 5 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MXSTDSETAN Supplier SPEX CertiPrep Group LLC Supplier No. MXSTDSETAN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Mixed Calibration Multi-Element Standards without Mercury for AA & ICP, 125 mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Mixed Calibration Standards
  • 125 mL Volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Mixed Multi-Element Calibration Standards without Mercury
Packaging Bottle
Name Note Without Mercury
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Assurance Grade Mixed Calibration Standard 1A,Assurance Grade Mixed Calibration Standard 2A,Assurance Grade Mixed Calibration Standard 3A
Technique Calibration of Conductivity Instrument
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 5% HNO3/Tr HF,5% HNO3,5% HNO3/tr HF
Quantity 5 x 125 mL
Grade Assurance
Show More Show Less

Product intended for laboratory use only