missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 4A For Environmental Analysis, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Mixed Calibration Standards
- Includes Mercury
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Mixed Calibration Standard 4A |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Nitric Acid/Trace Hydrogen Fluoride |
| Quantity | 500 mL |
| Grade | Environmental |
| Concentration or Composition (by Analyte or Components) | Components in 5% HNO3/tr. HF. Mercury is supplied separately. Al •100μg/mL Cr •50μg/mL Hg •100μg/mL SiO2 •100μg/mL Sn •40μg/mL Ti •100μg/mL Zn •50μg/mL |
Product intended for laboratory use only