missing translation for 'onlineSavingsMsg'
Learn More

Mixed Anion Standard, Ricca Chemical

Catalog No. ICMIX011A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICMIX011A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICMIX011A Supplier Ricca Chemical Company Supplier No. ICMIX011A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2000 ppm: Cl, SO4/500 ppm N (as NO3)/250 ppm Br/100 ppm F, N (as NO2), P (as PO4)

Specifications

Chemical Name or Material Mixed Anion Standard
Packaging Natural Poly Bottle
CAS 7722-76-1, 7732-18-5, 7757-79-1, 7647-14-5, 7757-82-6
CAS Min % 98.0, 0.06, 0.3, 0.3, 0.28, 0.03
CAS Max % 100.0, 0.07, 0.4, 0.35, 0.31, 0.05
Analyte or Component Names 100ppm F,N as NO2,N as NO3
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Multi-component Standard Solution
Element F, N as NO2, N as NO3
Quantity 1 L
Grade Laboratory
Show More Show Less