missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Mixed Anion Standard 7, Ricca Chemical

Catalog No. RV010738100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010738100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010738100 Supplier Ricca Chemical Company Supplier No. RV010738100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2 ppm F-, 10 ppm PO43-, NO3-, Br-, SO42-, 5ppm Cl-, NO2- /nManufactured and Tested in a

Specifications

Chemical Name or Material Verispec™ Mixed Anion Standard 7, 2 ppm F-, 10 ppm PO43-, NO3-, Br-, SO42-, 5 ppm Cl-, NO2-
Packaging Natural LDPE Bottle
Name Note Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
CAS 7681-49-4, 7732-18-5, 7758-02-3, 7783-20-2, 7763-00-0
CAS Min % 1, 0.0083, 0.0015, 0.0014, 0.0008, 0.0004
CAS Max % 1, 0.0083, 0.0015, 0.0014, 0.0008, 0.0004
Density 1.07
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Certified Reference Material
Quantity 100 mL
Grade High Purity
Show More Show Less