missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Long List Appendix IX Compounds Mix E, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC 8260EH
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 2-Butanone | 78-93-3 | 2000μg/mL |
| 2-Chloroethyl vinyl ether | 110-75-8 | 2000μg/mL |
| 2-Hexanone | 591-78-6 | 2000μg/mL |
| 4-Methyl-2-pentanone | 108-10-1 | 2000μg/mL |
| Acetone | 67-64-1 | 2000μg/mL |
| Carbon disulfide | 75-15-0 | 2000μg/mL |
| Vinyl acetate | 108-05-4 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Long List Appendix IX Mix E (High Level) | |
| 2mL amber prescored glass ampule | |
| For US EPA Method 8260B | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| Methanol-P and T | |
| 2000μg/mL |
| Method 8260B | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| AA, ICP | |
| Certified Reference Material | |
| 1 mL |
Product intended for laboratory use only