missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purgeable Gases, Mix B, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC 5022B
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Bromomethane | 74-83-9 | 200μg/mL |
| Chloroethane | 75-00-3 | 200μg/mL |
| Chloromethane | 74-87-3 | 200μg/mL |
| Dichlorodifluoromethane | 75-71-8 | 200μg/mL |
| Trichlorofluoromethane | 75-69-4 | 200μg/mL |
| Vinyl Chloride | 75-01-4 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Purgeable Gases, Mix B | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| AA, ICP | |
| Certified Reference Material | |
| 1 mL |
| 2mL amber prescored glass ampule | |
| For US EPA Methods 502.1, 502.2, 601, CLP | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| Methanol-P and T |
Product intended for laboratory use only