Learn More
Mitosis, Meiosis and Fission Set
Shows IPMAT in slides.
Supplier:  Customer Channels Group (Ccg) S10044
Catalog No. S10044
Only null left
Explore available promotions
Includes:
  • Vicia faba (broad bean) root tip squash, showing all stages of mitosis, stained with Feulgen reaction for DNA. Vicia has 12 large chromosomes
  • Whitefish mitosis, sections of Coregonus blasto-disc, all stages of mitosis shown
  • Lily meiosis, first meiotic divisions in cross section of lilium anther
  • Grasshopper Testis, l.s. to show spermatogenesis and spermiogenesis; stained with iron-hematoxylin
  • Paramecium caudatum in fission, w.m. of mass culture including several specimens in fission
Description
Description
Set of five slides shows the stages of nuclear division, cytokinesis, reduction division and even fission in Paramecium.
Specifications
Specifications
Mitosis Meiosis and Fission
5/Pk.
Product Suggestions
Product Suggestions