missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Fisher BioReagents™

Catalog No. p-27219
Change view
Click to view available options
Quantity:
2 mg
10 mg
20 mg
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25312 2 mg Amber Glass
BP253110 10 mg Amber Glass
BP253120 20 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP25312 Supplier Fisher BioReagents Supplier No. BP25312
Only null left

Mitomycin C

Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.

Recommended Storage:
4°C. Keep containers tightly closed in a dry, cool and well-ventilated place. Keep refrigerated.

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Melting Point 360°C
Color Blue
Assay Percent Range ≥98.0 %
Packaging Amber Glass
Quantity 2 mg
Merck Index 15, 6300
Solubility Information Soluble in water
Formula Weight 334.3
Percent Purity 99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10)
Physical Form Solid
Chemical Name or Material Mitomycin C
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Toxic if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is not required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:0
Recommended Storage 4°C