missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies Mito-rOCR Assay

The Agilent Mito-rOCR Assay makes mitochondrial function analysis easy and accessible. Measure relative oxygen consumption rate on a plate reader.

$3,484.65 - $7,939.15

Specifications

Detection Method Fluorescence
For Use With (Equipment) Synergy H1 Hybrid Multi-Mode Reader, Cytation 1 Cell Imaging Multi-Mode Reader, Cytation 5 Cell Imaging Multi-Mode Reader, Cytation 7 Cell Imaging Multi-Mode Reader, Cytation C10 Confocal Imaging Reader, Synergy Neo2 Hybrid Multi-Mode Microplate Reader
Cell Line Any Adherent Cell Type
System Type Mito-rOCR
Druggable Target Cell-based Assay
View More Specs

Products 5
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
MO4004
View Documents
Agilent Technologies
MO4004
Starter kit: reagents, plates, seal lids, magnetic holder and software (3 license seats) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MO3004
View Documents
Agilent Technologies
MO3004
Reagents, plates and seal lids for 4 assays N/A
Only null left
 
MO100
View Documents
Agilent Technologies
MO100
Magnetic holder only N/A
Only null left
 
MO2004
View Documents
Agilent Technologies
MO2004
Mito-rOCR microplates and seal lids only N/A
Only null left
 
MO500
View Documents
Agilent Technologies
MO500
Mito-rOCR Analysis Module software, 3 license seats N/A
Only null left
 
Description

Description

  • Simplifies mitochondrial respiration assessment with an intuitive, easy-to-use design.
  • Provides quick & reliable rOCR measurements for efficient data collection.
  • Eliminates the need for oil with Mito-rOCR seal lids, streamlining the assay process.
  • Compatible with a wide range of BioTek fluorescence plate readers for versatile use.
  • Glass-like plate bottom allows for data multiplexing and imaging of the same samples post-assay.
  • Automated data analysis module accelerates research with fast and robust results.
  • Reduces assay and analysis time by up to five times compared to traditional oil-based methods.
Specifications

Specifications

Fluorescence
Any Adherent Cell Type
Cell-based Assay
Synergy H1 Hybrid Multi-Mode Reader, Cytation 1 Cell Imaging Multi-Mode Reader, Cytation 5 Cell Imaging Multi-Mode Reader, Cytation 7 Cell Imaging Multi-Mode Reader, Cytation C10 Confocal Imaging Reader, Synergy Neo2 Hybrid Multi-Mode Microplate Reader
Mito-rOCR
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'