Learn More
Agilent Technologies Mito-rOCR Assay
The Agilent Mito-rOCR Assay makes mitochondrial function analysis easy and accessible. Measure relative oxygen consumption rate on a plate reader.
Supplier: Agilent Technologies MO3004
Description
- Simplifies mitochondrial respiration assessment with an intuitive, easy-to-use design.
- Provides quick & reliable rOCR measurements for efficient data collection.
- Eliminates the need for oil with Mito-rOCR seal lids, streamlining the assay process.
- Compatible with a wide range of BioTek fluorescence plate readers for versatile use.
- Glass-like plate bottom allows for data multiplexing and imaging of the same samples post-assay.
- Automated data analysis module accelerates research with fast and robust results.
- Reduces assay and analysis time by up to five times compared to traditional oil-based methods.
Specifications
| Fluorescence | |
| Reagents, plates and seal lids for 4 assays | |
| Cell-Based Assay Kit | |
| Mito-rOCR |
| Synergy H1 Hybrid Multi-Mode Reader, Cytation 1 Cell Imaging Multi-Mode Reader, Cytation 5 Cell Imaging Multi-Mode Reader, Cytation 7 Cell Imaging Multi-Mode Reader, Cytation C10 Confocal Imaging Reader, Synergy Neo2 Hybrid Multi-Mode Microplate Reader | |
| 4 Tests | |
| Any Adherent Cell Type | |
| Cell-based Assay |