missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ Ingenio™ Electroporation Kit

Catalog No. MIR50119
Change view
Click to view available options
Includes:
0.2 cm Cuvettes
0.4 cm Cuvettes
Quantity:
100 Reactions
25 Reactions
50 Reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
MIR50119 0.4 cm Cuvettes 100 Reactions
MIR50112 0.2 cm Cuvettes 25 Reactions
MIR50116 0.4 cm Cuvettes 50 Reactions
MIR50115 0.2 cm Cuvettes 50 Reactions
MIR50113 0.4 cm Cuvettes 25 Reactions
MIR50118 0.2 cm Cuvettes 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MIR50119 Supplier Mirus Bio™ Supplier No. MIR50119

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use the Ingenio™ Electroporation Kit for high efficiency electroporation using any instrument.

  • High efficiency electroporation of hard to transfect cell lines and primary cells
  • Compatible with most conventional electroporation devices including Lonza-Amaxa™, Bio-Rad™ or Harvard BTX™
  • Ideal for plasmid or siRNA electroporation

  • Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
  • Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
  • Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
  • High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes

Specifications

For Use With (Application) Plasmid or siRNA electroporation
For Use With (Equipment) Bio-Rad™ gene pulser and Harvard-BTX™ electroporator
Product Type Electroporation Kit
Includes 0.4 cm Cuvettes
Quantity 100 Reactions
Sterility Sterile
Content And Storage 4°C
Pulse Form Exponential Decay Wave