missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ Ingenio™ Electroporation Kit
Use the Ingenio™ Electroporation Kit for high efficiency electroporation using any instrument.
Supplier: Mirus Bio™ MIR 50113
Description
- High efficiency electroporation of hard to transfect cell lines and primary cells
- Compatible with most conventional electroporation devices including Lonza-Amaxa™, Bio-Rad™ or Harvard BTX™
- Ideal for plasmid or siRNA electroporation
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Chemical Identifiers
| Plasmid or siRNA electroporation | |
| Sterile | |
| Exponential Decay Wave |
| Electroporation Kit | |
| 4°C |
Specifications
| Plasmid or siRNA electroporation | |
| Electroporation Kit | |
| 25 Reactions | |
| 4°C |
| Bio-Rad™ gene pulser and Harvard-BTX™ electroporator | |
| 0.4 cm Cuvettes | |
| Sterile | |
| Exponential Decay Wave |