missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ TransIT-TKO™ siRNA Transfection Reagent
Description
- Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
- Low Cellular Toxicity: Maintain cell density and reduce experimental biases
- Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
- High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
- Concentration: 1.37 mg/mL
Specifications
Specifications
| Cell Type | Mammalian |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Concentration | 1.37 mg/mL |
| For Use With (Application) | Transfect small interfering RNA for knock out of target gene expression, optimal gene silencing in a large percentage of cells |
| Product Type | Transfection Reagent |
| Quantity | 0.4 mL |
| Sterility | Sterile |