missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT-TKO™ siRNA Transfection Reagent

Catalog No. MIR2154
Change view
Click to view available options
Quantity:
0.4 mL
5 x 1.5 mL
10 x 1.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2154 0.4 mL
MIR2155 5 x 1.5 mL
MIR2156 10 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MIR2154 Supplier Mirus Bio™ Supplier No. MIR 2154
Only null left

Enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types, including primary cells. siRNA and DNA co-transfection is feasible with TransIT-TKO. Transfections do not require medium changes and can be performed in serum-containing medium.

  • Broad Spectrum siRNA Delivery: Utilize one transfection reagent and protocol for a wide variety of cells
  • Low Cellular Toxicity: Maintain cell density and reduce experimental biases
  • Reproducible Results: Obtain consistent, targeted gene knockdowns from day to day
  • High Knockdown Efficiency: Achieve optimal gene silencing in a large percentage of cells to ensure experimental success
  • Concentration: 1.37 mg/mL

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 1.37 mg/mL
For Use With (Application) Transfect small interfering RNA for knock out of target gene expression, optimal gene silencing in a large percentage of cells
Product Type Transfection Reagent
Quantity 0.4 mL
Sterility Sterile