missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ TransIT™-LT1 Transfection Reagent
Description
- Broad spectrum DNA delivery: utilize one transfection reagent and protocol for a variety of cells
- Low cellular toxicity: maintain cell density and reduce experimental biases
- High efficiency delivery: achieve expression in a large population of cells for experimental success
- Deliver single or multiple plasmids: suitable for many applications such as gene expression, shRNA expression, viral production, and promoter analysis
Specifications
Specifications
| Content And Storage | Store for up to one year at 4°C. |
| Cell Type | Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | Gene expression, shRNA expression, viral production and promoter analysis |
| Product Type | Transfection Reagent |
| Quantity | 10 x 1 mL |
| Sterility | Sterile |