missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT™-LT1 Transfection Reagent

Catalog No. MIR2304
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2304 0.4 mL
MIR2300 1 mL
MIR2305 5 x 1 mL
MIR2306 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2304 Supplier Mirus Bio™ Supplier No. MIR 2304
Only null left

High-efficiency plasmid DNA delivery in many mammalian cell types, including primary cells. The TransIT™-LT1 Transfection Reagent is broad spectrum, low toxicity, and serum compatible, and is suitable for both transient and stable transfection.

  • Broad spectrum DNA delivery: utilize one transfection reagent and protocol for a variety of cells
  • Low cellular toxicity: maintain cell density and reduce experimental biases
  • High efficiency delivery: achieve expression in a large population of cells for experimental success
  • Deliver single or multiple plasmids: suitable for many applications such as gene expression, shRNA expression, viral production, and promoter analysis

Specifications

Content And Storage Store for up to one year at 4°C.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Gene expression, shRNA expression, viral production and promoter analysis
Product Type Transfection Reagent
Quantity 0.4 mL
Sterility Sterile