missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT™-LT1 Transfection Reagent

Catalog No. MIR2300
Encompass
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2300 1 mL
MIR2304 0.4 mL
MIR2305 5 x 1 mL
MIR2306 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2300 Supplier Mirus Bio™ Supplier No. MIR 2300
Only null left

High-efficiency plasmid DNA delivery in many mammalian cell types, including primary cells. The TransIT™-LT1 Transfection Reagent is broad spectrum, low toxicity, and serum compatible, and is suitable for both transient and stable transfection.

  • Broad spectrum DNA delivery: utilize one transfection reagent and protocol for a variety of cells
  • Low cellular toxicity: maintain cell density and reduce experimental biases
  • High efficiency delivery: achieve expression in a large population of cells for experimental success
  • Deliver single or multiple plasmids: suitable for many applications such as gene expression, shRNA expression, viral production, and promoter analysis

Specifications

Content And Storage Store for up to one year at 4°C.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Gene expression, shRNA expression, viral production and promoter analysis
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile