missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ Label IT™ siRNA Tracker™ Intracellular Localization Kit

Catalog No. MIR7212
Change view
Click to view available options
Product Type:
Cy3 Labeling Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
MIR7212 Cy3 Labeling Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR7212 Supplier Mirus Bio™ Supplier No. MIR7212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Label IT siRNA Tracker Reagent, Label IT Reconstitution Solution, and 10X Labeling Buffer A

Label, transfect, and track any siRNA residues in an efficient and nondestructive manner. LabelIT siRNA Tracker Intracellular Localization Kit provides a straight-forward approach to directly label siRNA of any sequence containing guanine residues.

  • Generate a labeled siRNA sample without hindering its inherent ability to silence targeted genes.
  • Monitor both sub-cellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells
  • Contain sufficient reagents to label 50μg of siRNA
  • siRNA can be delivered to cells in vitro or in vivo using the transfection reagent of your choice

Specifications

Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Vial
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection, Transient Transfection
For Use With (Application) Monitor both subcellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells
Includes Label IT™ siRNA tracker reagent, reconstitution solution, 10X Labeling Buffer A and siRNA dilution Buffer
Product Type Cy3 Labeling Kit
Quantity 1 Ea.
Sterility Sterile