missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ Label IT™ RNAi Delivery Controls
Description
- Sensitive - Easily visualize transfected cells and assess transfection efficiency using fluorescent microscopy
- Inert - Does not target any mammalian genes or cause off-target effects
- Compatible - Suitable for co-delivery experiments with functional siRNA
- Ease of Use - Supplied as a ready to use 10 μM stock solution with a 10X RNAi Dilution Buffer
- Visualization and tracking of transfected RNAi duplexes
Specifications
Specifications
| Format | Liquid |
| Product Type | Label IT RNAi Delivery Control |
| Stability | 6 Months |
| Storage Buffer | Dilution Buffer (50 mM Tris, pH 7.5, 100 mM NaCl) |
| Content And Storage | -20°C |
| Concentration | 7.5 nmol |
| Label or Dye | Cy™3 |
| Quantity | 100 μg |
| For Use With (Application) | in-vitro or in vivo tracking using direct fluorescent microscopy, co-delivery experiments with functional siRNA |