Learn More
Mirus Bio™ CHOgro™ Expression Media
Use CHOgro™ Expression Media for high density growth and quick adaptation of suspension CHO cells.
Supplier:  Mirus Bio™ MIR 6200
Catalog No. MIR6200
Only null left
Description
Description
CHOgro™ Expression Medium is a chemically defined, hydrolysate-free and animal-origin-free medium manufactured under cGMP conditions in an ISO-compliant facility. CHOgro™ is formulated to provide high density cell growth, and many suspension CHO cells (e.g. Freestyle CHO-S) can easily and quickly grow in CHOgro™ Expression Medium with minimal adaptation. CHOgro™ Expression Medium requires supplementation with L-Glutamine and Poloxamer 188 before use.
CHOgro™ Complex Formation Solution is an animal-origin-free solution specifically formulated for optimal transfection complex formation in conjunction with CHOgro™ Expression Media and TransIT-PRO™ Transfection Reagent to provide a completely animal-origin-free expression system.
  • High Density Growth – CHO-S cells grow > 12 x 106cells/mL
  • Quick Adaptation – CHO-S cells double within 24 hours of complete media change from FreeStyle™ Expression Media
  • No Cell Clumping Post-Transfection – Obtain accurate cell counts and high viability
  • Quality – CHOgro™ Medium and Complex Formation Solution are manufactured under cGMP conditions in an ISO-compliant facility
  • Regulatory Friendly – ALL components are animal origin free
Specifications
Specifications
CHO-derived
Liquid
Serum-free
Sterile-filtered
Ideal for Biotherapeutic Protein Production in Suspension CHO cells
Suspension Cell Culture
Chemically-defined, Animal Origin-free, Protein Production
Media
Sterile
Poloxamer 188, Glutamine
1 L
CHO
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : As labeled on product