missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirtazapine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 85650-52-8 |
|---|---|
| Molecular Formula | C17H19N3 |
| Molecular Weight (g/mol) | 265.35 |
| InChI Key | RONZAEMNMFQXRA-UHFFFAOYSA-N |
| Synonym | mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin |
| PubChem CID | 4205 |
| ChEBI | CHEBI:6950 |
| SMILES | CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC456170010
|
N/A | 1g |
N/A
|
|
|||||
|
AC456170050
|
N/A | 5g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 85650-52-8 | |
| 265.35 | |
| mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin | |
| CHEBI:6950 |
| C17H19N3 | |
| RONZAEMNMFQXRA-UHFFFAOYSA-N | |
| 4205 | |
| CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4 |
Specifications
| 85650-52-8 | |
| 97.5% min. (HPLC) | |
| C17H19N3 | |
| RONZAEMNMFQXRA-UHFFFAOYSA-N | |
| 265.35 | |
| CHEBI:6950 | |
| 98% |
| Authentic | |
| Glass Bottle | |
| mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin | |
| CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4 | |
| 4205 | |
| 265.35 | |
| Mirtazapine |
Safety and Handling
EINECSNumber : 288-060-6
RUO – Research Use Only