missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine, 98%

Catalog No. AC456170050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456170050 5 g
AC456170010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456170050 Supplier Thermo Scientific Chemicals Supplier No. 456170050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85650-52-8 | C17H19N3 | 265.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85650-52-8
Molecular Formula C17H19N3
Molecular Weight (g/mol) 265.35
InChI Key RONZAEMNMFQXRA-UHFFFAOYSA-N
Synonym mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin
PubChem CID 4205
ChEBI CHEBI:6950
SMILES CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4

Specifications

Melting Point 121.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 265.35
Percent Purity 98%
Chemical Name or Material Mirtazapine
EINECSNumber 288-060-6

RUO – Research Use Only