Learn More
PeproTech MIP-1 gamma Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: PeproTech 500P117BT500UG
Description
AA Sequence of recombinant protein: QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1gamma (CCL9/10). Anti-Murine MIP-1gamma (CCL9/10)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine MIP-1gamma (CCL9/10) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine MIP-1gamma (CCL9/10) (500-P117) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MIP-1gamma (CCL9/10). Western Blot: To detect Murine MIP-1gamma by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine MIP-1gamma is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P117BT-1MG will be provided as 2 x 500 μg
MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.Chemical Identifiers
| MIP-1 gamma | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Mouse | |
| P51670 | |
| 20308 | |
| E.coli-derived Recombinant Murine MIP-1gamma (CCL9/10). | |
| Antigen affinity chromatography | |
| Ccl9 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9 | |
| Ccl9 | |
| Primary | |
| -20°C |
Specifications
| MIP-1 gamma | |
| Polyclonal | |
| Biotin | |
| Ccl9 | |
| C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20308 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P51670 | |
| Ccl9 | |
| E.coli-derived Recombinant Murine MIP-1gamma (CCL9/10). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |