missing translation for 'onlineSavingsMsg'
Learn More

MIP-1 delta Monoclonal Antibody (K70043_3C5), Invitrogen™

Mouse Monoclonal Antibody

$1557.73 - $2745.56

Specifications

Antigen MIP-1 delta
Clone K70043_3C5
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555662
View Documents
Invitrogen™
MA555662
50 μL
Each for $1,557.73
Only null left
 
MA556042
View Documents
Invitrogen™
MA556042
200 μL
Each for $2,745.56
Only null left
 
Description

Description

Sequence of this protein is as follows: QFTNDAETEL MMSKLPLENP VVLNSFHFAA DCCTSYISQS IPCSLMKSYF ETSSECSKPG VIFLTKKGRQ VCAKPSGPGV QDCMKKLKPY SI

MIP-1 delta Monoclonal antibody specifically detects MIP-1 delta in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

MIP-1 delta
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2Lkn-1; HMRP-2B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1D; MIP5; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC3; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15
Recombinant human CCL15 (immunogen range is 22-113).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CCL15
K70043_3C5
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Rat
Q16663
6359
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'