missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MIP-1 beta ProQuantum Immunoassay Kit
ELISA
$990.00 - $4150.00
Specifications
| Accession Number | P13236 |
|---|---|
| Assay Range | 0.064 to 5000 pg/mL |
| Assay Sensitivity | <0.05 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
Description
CCL4 (macrophage inflammatory protein 1-beta, MIP1B) belongs to the intercrine beta (chemokine CC) family. Functionally, CCL4 is involved in chemotactic and proinflammatory effects, and homeostasis. CCL4 is produced by macrophages upon stimulation by bacterial endotoxins. CCL4 recruits and stimulates various inflammatory cells at sites of inflammation. CCL4 is produced by lymphocytes, macrophages and dendritic cells. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells. CCL4 binds with high affinity to CCR5 receptors.Specifications
| P13236 | |
| <0.05 pg/mL | |
| Taqman™ Real-time PCR | |
| ProQuantum | |
| Human | |
| qPCR Instruments | |
| ACT2; AT744.1; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP1B; MIP1B1; SCYA2; SCYA4 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| RUO | |
| Frozen at -20°C | |
| 30 mins |
| 0.064 to 5000 pg/mL | |
| FAM™ | |
| Proximity-based Immunoassay | |
| Plasma, Serum, Supernatant | |
| Protein Quantitation | |
| 6351 | |
| 1 hr. | |
| FAM™ | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| CCL4 | |
| 2 hrs |