Learn More
Minuteman™ Vacuum Accessories and Replacement Parts
$8.67 - $468.73
Specifications
For Use With (Application) Remediation of Hazardous Molds, Spores, Bio-haz Dust and Viral Particulate Containment
View More Specs
Products 4
Catalog Number Mfr. No. Product Type For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type For Use With (Equipment) Quantity Price Quantity & Availability  
18999970
Encompass
View Documents Promotion Details
Minuteman™
110029
ULPA Replacement Filter Bio-haz 6 gal. and 15 gal. and X-829 6 gal. and 15 gal., and Lead vacuum Each
Each for $468.73
Only null left
19063053
Encompass
View Documents Promotion Details
Minuteman™
800056
Vacuum Filter Bio-haz. 6 gal., X-829 6 gal., Lead Vacuum, X-839 Each
Each for $175.32
Only null left
19163074
Encompass
View Documents Promotion Details
Minuteman™
802206PKG
Encapsulation Bag Minuteman Bio-haz Vacuum (6 gal.) 10 Pk.
Pack of 10 Each for $8.67
Only null left
18999991
Encompass
View Documents Promotion Details
Minuteman™
805038PKG
Filter Protector Bio-haz 15 gal., X-829 15 gal. units, X-839 Series, X-250/X-380, X-1000-15 and MX-1000-15, MRS-1,3,4, HEPA Filter Vacuum , X702-55, 290 Series 15/20 gal., 390 Series 15 gal., 390 Twin Series 10 Pk.
Pack of 12 Each for $82.27
Only null left
Description
Description
Impact filter can be used with both tank sizes. All other replacement parts are size specific. Plastic biohazard encapsulation bags assist in proper collection and disposal of potentially hazardous contaminants.
Specifications
Specifications
Remediation of Hazardous Molds, Spores, Bio-haz Dust and Viral Particulate Containment