Learn More
Minuteman™ Vacuum Accessories and Replacement Parts
Catalog No. 19163074
Encompass
Change view
Click to view available options
Product Type:
Encapsulation Bag
Filter Protector
ULPA Replacement Filter
Vacuum Filter
For Use With (Equipment):
Bio-haz 15 gal., X-829 15 gal. units, X-839 Series, X-250/X-380, X-1000-15 and MX-1000-15, MRS-1,3,4, HEPA Filter Vacuum , X702-55, 290 Series 15/20 gal., 390 Series 15 gal., 390 Twin Series
Bio-haz 6 gal. and 15 gal. and X-829 6 gal. and 15 gal., and Lead vacuum
Bio-haz. 6 gal., X-829 6 gal., Lead Vacuum, X-839
Minuteman Bio-haz Vacuum (6 gal.)
Quantity:
10 Pk.
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment) Quantity
19163074 Encapsulation Bag Minuteman Bio-haz Vacuum (6 gal.) 10 Pk.
18999970 ULPA Replacement Filter Bio-haz 6 gal. and 15 gal. and X-829 6 gal. and 15 gal., and Lead vacuum Each
19063053 Vacuum Filter Bio-haz. 6 gal., X-829 6 gal., Lead Vacuum, X-839 Each
18999991 Filter Protector Bio-haz 15 gal., X-829 15 gal. units, X-839 Series, X-250/X-380, X-1000-15 and MX-1000-15, MRS-1,3,4, HEPA Filter Vacuum , X702-55, 290 Series 15/20 gal., 390 Series 15 gal., 390 Twin Series 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19163074 Supplier Minuteman™ Supplier No. 802206PKG
Impact filter can be used with both tank sizes. All other replacement parts are size specific. Plastic biohazard encapsulation bags assist in proper collection and disposal of potentially hazardous contaminants.
Specifications
For Use With (Application) Remediation of Hazardous Molds, Spores, Bio-haz Dust and Viral Particulate Containment
Product Type Encapsulation Bag
Quantity 10 Pk.
For Use With (Equipment) Minuteman Bio-haz Vacuum (6 gal.)
Material Plastic