missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil

Catalog No. AAJ6180303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6180303 1 g
AAJ6180306 5 g
AAJ6180314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6180303 Supplier Thermo Scientific Chemicals Supplier No. J6180303

Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels. | CAS: 38304-91-5 | C9H15N5O | 209.253 g/mol

Minoxidil is used as antihypertensive, antialopecia agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
  • Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.253
MDL Number MFCD00063409
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
Synonym minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
PubChem CID 4201
IUPAC Name 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O

Specifications

Melting Point 248°C to 265°C
Color White
Quantity 1 g
Merck Index 14,6203
Solubility Information Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol.
Formula Weight 209.25
Physical Form Crystalline Powder
Chemical Name or Material Minoxidil
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 253-874-2
RTECSNumber UV8200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only