missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minoxidil 98.0+%, TCI America™
$57.79 - $162.17
Chemical Identifiers
| CAS | 38304-91-5 |
|---|---|
| Molecular Formula | C9H15N5O |
| Molecular Weight (g/mol) | 209.253 |
| MDL Number | MFCD00063409 |
| InChI Key | ZIMGGGWCDYVHOY-UHFFFAOYSA-N |
| Synonym | minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox |
| PubChem CID | 4201 |
| IUPAC Name | 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine |
| SMILES | C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O |
Chemical Identifiers
| 38304-91-5 | |
| 209.253 | |
| ZIMGGGWCDYVHOY-UHFFFAOYSA-N | |
| 4201 | |
| C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O |
| C9H15N5O | |
| MFCD00063409 | |
| minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox | |
| 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine |
Specifications
| 38304-91-5 | |
| C9H15N5O | |
| 1 g | |
| ZIMGGGWCDYVHOY-UHFFFAOYSA-N | |
| 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine | |
| 4201 | |
| ≥98.0% (HPLC,T) | |
| Minoxidil |
| White-Yellow | |
| MFCD00063409 | |
| minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox | |
| C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O | |
| 209.253 | |
| 209.25 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UV8200000
TSCA : No