missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil 98.0+%, TCI America™

Catalog No. M13891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13891G 1 g
M13895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13891G Supplier TCI America Supplier No. M13891G

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.253
MDL Number MFCD00063409
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
Synonym minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
PubChem CID 4201
IUPAC Name 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 209.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Minoxidil
RTECSNumber UV8200000
TSCA No