missing translation for 'onlineSavingsMsg'
Learn More

Minodronate Monohydrate 98.0+%, TCI America™

Catalog No. M236025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M236025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M236025MG Supplier TCI America Supplier No. M236025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 155648-60-5
Molecular Formula C9H14N2O8P2
Molecular Weight (g/mol) 340.165
MDL Number MFCD00890772
InChI Key GPAPAOGRNKUFGH-UHFFFAOYSA-N
Synonym 1-Hydroxy-2-[imidazo[1,2-a]pyridin-3-yl]ethylidene-1,1-diphosphonic Acid, Minodronic Acid
PubChem CID 23201029
IUPAC Name (1-hydroxy-2-imidazo[1,2-a]pyridin-3-yl-1-phosphonoethyl)phosphonic acid;hydrate
SMILES C1=CC2=NC=C(N2C=C1)CC(O)(P(=O)(O)O)P(=O)(O)O.O

Specifications

Color White
Quantity 25 mg
UN Number 2811
Formula Weight 322.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Minodronate Monohydrate
RTECSNumber SZ8562470
TSCA No