accessibility menu, dialog, popup

UserName

CAS RN 155648-60-5

IUPAC Name: (1-hydroxy-2-{imidazo[1,2-a]pyridin-3-yl}-1-phosphonoethyl)phosphonic acid hydrate
Synonyms: minodronic acid hydrate
Molecular Weight (g/mol): 340.17
Molecular Formula: C9H14N2O8P2
InChi Key: GPAPAOGRNKUFGH-UHFFFAOYSA-N
SMILES: O.OC(CC1=CN=C2C=CC=CN12)(P(O)(O)=O)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results