missing translation for 'onlineSavingsMsg'
Learn More

Minocycline Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America M22881G

 View more versions of this product

Catalog No. M22881G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Minocycline Hydrochloride
Yellow
MFCD00083669
minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
493.94
493.94
Crystalline Powder
13614-98-7
C23H28ClN3O7
1 g
KDLQIOPKJDNQIM-WUURTAMISA-N
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
54685925
≥98.0% (HPLC,T)

Chemical Identifiers

13614-98-7
493.94
KDLQIOPKJDNQIM-WUURTAMISA-N
54685925
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
C23H28ClN3O7
MFCD00083669
minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QI7630500

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions