missing translation for 'onlineSavingsMsg'
Learn More

Minocycline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133138
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22881G 1 g
M22885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22881G Supplier TCI America Supplier No. M22881G
Only null left

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
Synonym minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino
PubChem CID 54685925
IUPAC Name hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 493.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Minocycline Hydrochloride
RTECSNumber QI7630500
TSCA No
Recommended Storage Refrigerator