Learn More
Minigrip™ UV Guard™ UV Protection Bags
Prevent light-sensitive products from deterioration
Supplier:  Minigrip™ MGUV3P0608
 View more versions of this product
Catalog No. 22311212
Only null left
Description
Description
  • Amber
  • With Color Line™ and premium zipper
  • Film contains UV blocker
  • Thickness of 3 mil
  • Available in different sizes
Compliance
USP XXII and USP XXIII procedure 66.1.
Specifications
Chemical Identifiers
Premium Zipper
Polyethylene
UV Protection Bag
Amber
3 mil
Specifications
Premium Zipper
Polyethylene
6 x 8 in. (15.24 x 20.32cm)
8 in.
Amber
3 mil
6 in.
UV Protection Bag
Product Suggestions
Product Suggestions