Learn More
Minigrip™ UV Guard™ UV Protection Bags
Prevent light-sensitive products from deterioration
Supplier:  Minigrip™ MGUV3P0203
 View more versions of this product
Catalog No. 22311207
Only null left
Description
Description
  • Amber
  • With Color Line™ and premium zipper
  • Film contains UV blocker
  • Thickness of 3 mil
  • Available in different sizes
Compliance
USP XXII and USP XXIII procedure 66.1.
Specifications
Chemical Identifiers
Premium Zipper
Polyethylene
UV Protection Bag
Amber
3 mil
Specifications
Premium Zipper
Polyethylene
2 x 3 in. (5.08 x 7.62cm)
3 in.
Amber
3 mil
2 in.
UV Protection Bag
Product Suggestions
Product Suggestions