missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minigrip™ Premium Red Line™ Reclosable Zipper Bags
Many size and film thickness options
Supplier: Minigrip™ MGRL2P1212
Description
- Premium zipper easily snaps open and zips securely shut
- Superior clarity that you can see
- Most sizes are packaged in convenient DispenserBag™ packaging
- Packaged in high quality corrugated boxes for reliable storage and easy handling
- Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
- Reinforced zipper and side seams to keep bags from splitting when opened or filled
Chemical Identifiers
| Zip Seal | |
| Clear | |
| LDPE, LLDPE | |
| 2 mil |
Specifications
| Zip Seal | |
| LDPE, LLDPE | |
| 12 in. |
| Clear | |
| 2 mil | |
| 12 in. |