Learn More
Minigrip™ Premium Red Line™ Reclosable Zipper Bags
Many size and film thickness options
Supplier:  Minigrip™ MGRL2P0608
 View more versions of this product
Catalog No. 22306361
Only null left
Description
Description
  • Premium zipper easily snaps open and zips securely shut
  • Superior clarity that you can see
  • Most sizes are packaged in convenient DispenserBag™ packaging
  • Packaged in high quality corrugated boxes for reliable storage and easy handling
  • Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
  • Reinforced zipper and side seams to keep bags from splitting when opened or filled
Specifications
Chemical Identifiers
Zip Seal
Clear
LDPE, LLDPE
2 mil
Specifications
Zip Seal
LDPE, LLDPE
8 in.
Clear
2 mil
6 in.
Product Suggestions
Product Suggestions