Learn More
Cytiva Mini Dialysis Kits
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
Supplier: Cytiva 80648413
Includes: 50 tubes with 50 dialysis and 50 standard caps, plus six floats
Description
- Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
- Uses disposable dialysis tubes with dialysis membrane incorporated into caps
- Conical tube bottom maximizes sample recovery
- Process complete in as little as several hours
- Choice of molecular weight cut-off and tube size
Specifications
| Mini Dialysis Kit | |
| 250 μL | |
| Each |
| 8 kDa | |
| 50 x tube with dialysis cap, 50 x standard cap, 6 x float | |
| Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |