missing translation for 'onlineSavingsMsg'
Learn More

Mineral oil, pure

Catalog No. p-4323678
Change view
Click to view available options
Quantity:
6 mL
1 L
2.5 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415080060 6 mL
AC415080010 1 L
AC415080025 2.5 L
AC415080100 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC415080060 Supplier Thermo Scientific Chemicals Supplier No. 415080060

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Density 0.8770g/mL
Flash Point 220°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4760 to 1.4830
Quantity 6 mL
Specific Gravity 0.877
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether, cs2,, petroleum ether, oils, miscible with wax and fats, insoluble in ethanol
Viscosity 65-75 mm2/s (40°C)
Grade Pure
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Show More Show Less
EINECSNumber 232-455-8

RUO – Research Use Only