missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Medium and Low Viscosity, FCC, Spectrum™ Chemical

Catalog No. 18604614
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604614 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604614 Supplier Spectrum Chemical Mfg Cor Supplier No. M11931LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1193, 8042-47-5

Spectrum™ Chemical Mineral Oil, Medium and Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8042-47-5

Specifications

CAS Max % 1
Boiling Point >230°C
Packaging Amber Glass Bottle
Quantity 1 L
Viscosity 3-11 cSt
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid