missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Medium and Low Viscosity, FCC, Spectrum™ Chemical

Catalog No. 18604617
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604617 20 L Amber Glass Bottle
18604613 500 mL Amber Glass Bottle
18604615 4 L Amber Glass Bottle
18604616 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604617 Supplier Spectrum Chemical Mfg Cor Supplier No. M119320LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1193, 8042-47-5

Spectrum™ Chemical Mineral Oil, Medium and Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8042-47-5

Specifications

CAS Max % 1
Boiling Point >230°C / 446°F
Packaging Amber Glass Bottle
Quantity 20 L
Viscosity 3-11 cSt
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Mineral Oil, Low Viscosity